m-Toluic acid ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
3-Methylbenzoic acid
T0291-500G
T0291-25G
TCT0291-500GEA
150.1
CAD
TCT0291-500G
TCT0291-025G
m-Toluic acid ≥98.0%
m-Toluic acid
CAS Number: 99-04-7
MDL Number: MFCD00002523
Molecular Formula: C8H8O2
Molecular Weight: 136.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 106
Melting point (°C): 112
Flash Point (°C): 150
Other Product Names / Synonyms: 3-Methylbenzoic Acid
Formula:
C₈H₈O₂ Boiling Pt: 106 °C Melting Pt: 112 °C Density: 1.054 g/cm³ (20 °C) Flash Pt: 150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002523 CAS Number: 99-04-7 EINECS: 202-723-9 Merck Index: 12,09673 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...