Thianthrene ≥98.0%

Supplier: TCI America

T0184-25G T0184-5G
TCT0184-25GEA 129.58 CAD
TCT0184-25G TCT0184-005G
Thianthrene ≥98.0%
Thianthrene

CAS Number: 92-85-3
MDL Number: MFCD00005065
Molecular Formula: C12H8S2
Molecular Weight: 216.32
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 156

Formula: C₁₂H₈S₂
Melting Pt: 156 °C
MDL Number: MFCD00005065
CAS Number: 92-85-3
EINECS: 202-197-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f06214e410d7ac49ad77aabc1394cdf1bef8e91&vid=6f06214e410d7ac49ad77aabc1394cdf1bef8e91&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR