N,N,N',N'-Tetramethylbenzidine 98,0

Supplier: TCI America

Synonyms: N,n,n,n-tetramethylbenzidine, 4,4'-Bis(dimethylamino)biphenyl

T0141-5G T0141-1G
TCT0141-5GEA 443.03 CAD
TCT0141-5G TCT0141-001G
N,N,N',N'-Tetramethylbenzidine 98,0
N,N,N',N'-Tetramethylbenzidine

CAS Number: 366-29-0
MDL Number: MFCD00008310
Molecular Formula: C16H20N2
Molecular Weight: 240.35
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 193


Other Product Names / Synonyms: 4,4'-Bis(dimethylamino)biphenyl

Formula: C₁₆H₂₀N₂
Melting Pt: 193 °C
MDL Number: MFCD00008310
CAS Number: 366-29-0
EINECS: 206-676-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c06488e9b85fb3cebd9899ef29cb2c476a16c2a7&vid=c06488e9b85fb3cebd9899ef29cb2c476a16c2a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR