Tetraethylammonium iodide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T0097-25G T0097-500G T0097-100G
TCT0097-025GEA 62.68 CAD
TCT0097-025G TCT0097-500G TCT0097-100G
Tetraethylammonium iodide ≥98.0% (by titrimetric analysis)
Tetraethylammonium iodide

CAS Number: 68-05-3
MDL Number: MFCD00011829
Molecular Formula: C8H20IN
Molecular Weight: 257.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White

Formula: C₈H₂₀IN
Melting Pt: 300 °C
Density: 1.566 g/cm³ (20 °C)
MDL Number: MFCD00011829
CAS Number: 68-05-3
EINECS: 200-676-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47afa75d64b86c8ce966b011c7375a8cd9c7bacb&vid=47afa75d64b86c8ce966b011c7375a8cd9c7bacb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR