Tetrachlorohydroquinone ≥98.0%

Supplier: TCI America

T0067-25G T0067-5G
TCT0067-025GEA 262.28 CAD
TCT0067-025G TCT0067-5G
Tetrachlorohydroquinone ≥98.0%
Tetrachlorohydroquinone

CAS Number: 87-87-6
MDL Number: MFCD00041748
Molecular Formula: C6H2Cl4O2
Molecular Weight: 247.88
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 242


Other Product Names / Synonyms: 1,2,4,5-Tetrachloro-3,6-dihydroxybenzene, Tetrachloroquinol

Formula: C₆H₂Cl₄O₂
Melting Pt: 242 °C
MDL Number: MFCD00041748
CAS Number: 87-87-6
EINECS: 201-779-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=332efe82ac95eb3968b12001dad35a178c0ef7e4&vid=332efe82ac95eb3968b12001dad35a178c0ef7e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR