Salicylaldehyde ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
S0275-25G
S0275-500G
TCS0275-025GEA
97.42
CAD
TCS0275-025G
TCS0275-500G
Salicylaldehyde ≥98.0% (by GC, titration analysis)
Salicylaldehyde
CAS Number: 90-02-8
MDL Number: MFCD00003317
Molecular Formula: C7H6O2
Molecular Weight: 122.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 196
Melting point (°C): 2
Flash Point (°C): 90
Specific Gravity (20/20): 1.17
Other Product Names / Synonyms: 2-Hydroxybenzaldehyde
Formula:
C₇H₆O₂ Boiling Pt: 196 °C Melting Pt: 2 °C Density: 1.17 (20 °C) Flash Pt: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003317 CAS Number: 90-02-8 EINECS: 201-961-0 Merck Index: 13,08405 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...