Lithium stearate ≥90.0%

Supplier: TCI America

Synonyms: Stearic acid lithium salt, Lithium Stearate

S0237-500G S0237-25G
TCS0237-500GEA 146.02 CAD
TCS0237-500G TCS0237-025G
Lithium stearate ≥90.0%
Lithium stearate

CAS Number: 4485-12-5
MDL Number: MFCD00042032
Molecular Formula: C18H36O2
Molecular Weight: 290.42
Purity/Analysis Method: >90.0% (T)
Form: Crystal
Color: White
Melting point (°C): 220


Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Stearic Acid Lithium Salt

Formula: C₁₈H₃₅LiO₂
Melting Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00042032
CAS Number: 4485-12-5
EINECS: 224-772-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d3b455d4eaa3a26caa0869a8d46d8e96a68cafb&vid=0d3b455d4eaa3a26caa0869a8d46d8e96a68cafb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR