Stearic anhydride ≥97.0%

Supplier: TCI America
Warning

S0083-25G S0083-500G
TCS0083-025GEA 167.48 CAD
TCS0083-025G TCS0083-500G
Stearic anhydride ≥97.0%
Stearic anhydride

CAS Number: 638-08-4
MDL Number: MFCD00009004
Molecular Formula: C36H70O3
Molecular Weight: 550.95
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Melting point (°C): 71

Formula: C₃₆H₇₀O₃
Melting Pt: 71 °C
Density: 0.855 g/cm³ (60 °C)
Storage Temperature: Freezer
MDL Number: MFCD00009004
CAS Number: 638-08-4
EINECS: 211-318-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31df99206f84dd2a5991b1d62fcd260c22d3fc38&vid=31df99206f84dd2a5991b1d62fcd260c22d3fc38&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR