Diethyl sebacate ≥98.0%

Supplier: TCI America

Synonyms: Sebacic acid diethyl ester, Decanedioic acid diethyl ester, Diethyl decanedioate

S0024-25ML S0024-500ML
TCS0024-025MLEA 44.24 CAD
TCS0024-025ML TCS0024-500ML
Diethyl sebacate ≥98.0%
Diethyl sebacate

CAS Number: 110-40-7
MDL Number: MFCD00009219
Molecular Formula: C14H26O4
Molecular Weight: 258.36
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 309
Flash Point (°C): 167
Specific Gravity (20/20): 0.97


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Decanedioic Acid Diethyl Ester, Sebacic Acid Diethyl Ester

Formula: C₁₄H₂₆O₄
MW: 258.36 g/mol
Boiling Pt: 309 °C
Melting Pt: –3 °C
Density: 0.97 (20 °C)
Flash Pt: 167 °C
Storage Temperature: Ambient
MDL Number: MFCD00009219
CAS Number: 110-40-7
EINECS: 203-764-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84d4d69f39338acaf5f3d6b7a7b78a2ef2ea9d82&vid=84d4d69f39338acaf5f3d6b7a7b78a2ef2ea9d82&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR