Quinaldine red ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Q0007-25G Q0007-1G
TCQ0007-025GEA 313.91 CAD
TCQ0007-025G TCQ0007-001G
Quinaldine red ≥95.0% (by titrimetric analysis)
Quinaldine red

CAS Number: 117-92-0
MDL Number: MFCD00011968
Molecular Formula: C21H23IN2
Molecular Weight: 430.33
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: Deep Blue

Formula: C₂₁H₂₃IN₂
Melting Pt: 240 °C
MDL Number: MFCD00011968
CAS Number: 117-92-0
EINECS: 204-221-5
Merck Index: 12,08230
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a759494a820b6172f4c5ca4d511a7f02a77b6608&vid=a759494a820b6172f4c5ca4d511a7f02a77b6608&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR