Quinaldic acid ≥98.0%

Supplier: TCI America
Warning

Synonyms: Quinoline-2-carboxylic acid, 2-Quinolinecarboxylic acid

Q0003-5G Q0003-25G
TCQ0003-005GEA 26.86 CAD
TCQ0003-005G TCQ0003-025G
Quinaldic acid ≥98.0%
Quinaldic acid

CAS Number: 93-10-7
MDL Number: MFCD00006752
Molecular Formula: C10H7NO2
Molecular Weight: 173.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 157


Other Product Names / Synonyms: 2-Quinolinecarboxylic Acid

Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Melting Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00006752
CAS Number: 93-10-7
EINECS: 202-218-3
Merck Index: 12,08227
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6542e939f45e06dc93afbad4dd2370e2c12c08ea&vid=ef0fdf83683e7d175c17408b86a702270540ad37&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR