1,1,1,2,2-Pentafluoro-4-iodobutane ≥98.0%

Supplier: TCI America

Synonyms: 1-Iodo-3,3,4,4,4-pentafluorobutane

P1817-5G P1817-25G
TCP1817-5GEA 93.22 CAD
TCP1817-5G TCP1817-25G
1,1,1,2,2-Pentafluoro-4-iodobutane ≥98.0%
1,1,1,2,2-Pentafluoro-4-iodobutane

CAS Number: 40723-80-6
MDL Number: MFCD00155910
Molecular Formula: C4H4F5I
Molecular Weight: 273.97
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 100
Specific Gravity (20/20): 1.93


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3,3,4,4,4-Pentafluorobutyl Iodide

Formula: C₄H₄F₅I
MW: 273.97 g/mol
Boiling Pt: 100 °C
Density: 1.93 (20 °C)
MDL Number: MFCD00155910
CAS Number: 40723-80-6
EINECS: 255-055-5
UN: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae32ca0becad1d787b20210a666755343522c048&vid=b93cd42d044775e9c9ab51189cc5df7d4dcfdf43&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR