Potassium (2-methoxyphenyl)trifluoroborate ≥98.0% (by HPLC)

Supplier: TCI America

P1810-1G P1810-5G
TCP1810-1GEA 135.88 CAD
TCP1810-1G TCP1810-5G
Potassium (2-methoxyphenyl)trifluoroborate ≥98.0% (by HPLC)
Potassium (2-methoxyphenyl)trifluoroborate

CAS Number: 236388-46-8
MDL Number: MFCD04115756
Molecular Formula: C7H7BF3KO
Molecular Weight: 214.04
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White

Formula: C₇H₇BF₃KO
MDL Number: MFCD04115756
CAS Number: 236388-46-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=130eecf3fe9acb88622d5b135775ba21436a2b53&vid=130eecf3fe9acb88622d5b135775ba21436a2b53&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR