Potassium(bromomethyl)trifluoroborate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

P1807-1G P1807-5G
TCP1807-1GEA 56.88 CAD
TCP1807-1G TCP1807-5G
Potassium(bromomethyl)trifluoroborate ≥98.0% (by titrimetric analysis)
Potassium (bromomethyl)trifluoroborate

CAS Number: 888711-44-2
MDL Number: MFCD09265154
Molecular Formula: CH2BBrF3K
Molecular Weight: 200.83
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 220

Formula: CH₂BBrF₃K
Melting Pt: 220 °C
MDL Number: MFCD09265154
CAS Number: 888711-44-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efe35f3988da87f915267828ee1207ce86d2c5ec&vid=efe35f3988da87f915267828ee1207ce86d2c5ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR