1-Phenyloxindole ≥98.0%
Supplier: TCI America
P1769-25G
P1769-5G
TCP1769-25GEA
82.16
CAD
TCP1769-25G
TCP1769-5G
1-Phenyloxindole ≥98.0%
1-Phenyloxindole
CAS Number: 3335-98-6
MDL Number: MFCD00234850
Molecular Formula: C14H11NO
Molecular Weight: 209.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 122
Other Product Names / Synonyms: 1-Phenyl-2-indolinone, 2-Oxo-1-phenylindoline
Formula:
C₁₄H₁₁NO Melting Pt: 122 °C |
MDL Number:
MFCD00234850 CAS Number: 3335-98-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...