Potassium(3-methoxyphenyl)trifluoroborate ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Potassium trifluoro(3-methoxyphenyl)borate, (3-Methoxyphenyl)trifluoroboric acid potassium salt

P1580-1G P1580-5G
TCP1580-1GEA 107.44 CAD
TCP1580-1G TCP1580-5G
Potassium(3-methoxyphenyl)trifluoroborate ≥98.0% (by HPLC)
Potassium (3-methoxyphenyl)trifluoroborate

CAS Number: 438553-44-7
MDL Number: MFCD04039954
Molecular Formula: C7H7BF3KO
Molecular Weight: 214.04
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 180

Formula: C₇H₇BF₃KO
MW: 214.04 g/mol
Melting Pt: 180 °C
MDL Number: MFCD04039954
CAS Number: 438553-44-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb1b060f5f43a4ef149199862ba72c4949c1563e&vid=cb1b060f5f43a4ef149199862ba72c4949c1563e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR