1-Phenyl-2-(trimethylsilyl)acetylene ≥98.0%

Supplier: TCI America

Synonyms: Trimethyl(phenylethynyl)silane

P1398-5ML P1398-25ML
TCP1398-5MLEA 71.1 CAD
TCP1398-5ML 76220-210
1-Phenyl-2-(trimethylsilyl)acetylene ≥98.0%
1-Phenyl-2-(trimethylsilyl)acetylene

CAS Number: 2170-06-1
MDL Number: MFCD00054860
Molecular Formula: C11H14Si
Molecular Weight: 174.32
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 88
Flash Point (°C): 73
Specific Gravity (20/20): 0.90


Other Product Names / Synonyms: (Phenylethynyl)trimethylsilane

Formula: C₁₁H₁₄Si
Boiling Pt: 88 °C
Density: 0.90 (20 °C)
Flash Pt: 73 °C
MDL Number: MFCD00054860
CAS Number: 2170-06-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6d59e659c2441a2cedb2e2af56ff132d978e546&vid=b6d59e659c2441a2cedb2e2af56ff132d978e546&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR