2-Phenylthiophene ≥95.0%

Supplier: TCI America

P1397-1G P1397-5G
TCP1397-1GEA 72.35 CAD
TCP1397-1G TCP1397-5G
2-Phenylthiophene ≥95.0%
2-Phenylthiophene

CAS Number: 825-55-8
MDL Number: MFCD00130080
Molecular Formula: C10H8S
Molecular Weight: 160.23
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 36
Storage Temperature: 0-10°C

Formula: C₁₀H₈S
MW: 160.24 g/mol
Melting Pt: 36 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00130080
CAS Number: 825-55-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4dab062729ca7725681e42617027f5ef0b70444&vid=c4dab062729ca7725681e42617027f5ef0b70444&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR