(5-Bromopentyl)benzene ≥98.0%
Supplier: TCI America
P1323-5G
P1323-25G
TCP1323-005GEA
278.08
CAD
TCP1323-005G
TCP1323-25G
(5-Bromopentyl)benzene ≥98.0%
(5-Bromopentyl)benzene
CAS Number: 14469-83-1
MDL Number: MFCD01075177
Molecular Formula: C11H15Br
Molecular Weight: 227.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 148
Specific Gravity (20/20): 1.23
Other Product Names / Synonyms: 5-Phenylpentyl Bromide
Formula:
C₁₁H₁₅Br Boiling Pt: 148 °C Density: 1.23 (20 °C) |
MDL Number:
MFCD01075177 CAS Number: 14469-83-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...