2-Phenyl-1,3-propanediol ≥98.0%

Supplier: TCI America
Danger

P1249-5G P1249-25G
TCP1249-005GEA 159.17 CAD
TCP1249-005G TCP1249-25G
2-Phenyl-1,3-propanediol ≥98.0%
2-Phenyl-1,3-propanediol

CAS Number: 1570-95-2
MDL Number: MFCD00236056
Molecular Formula: C9H12O2
Molecular Weight: 152.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 52

Formula: HOH₂CCH(C₆H₅)CH₂OH
Melting Pt: 52 °C
MDL Number: MFCD00236056
CAS Number: 1570-95-2
EINECS: 411-810-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7373bcd667fba9afe3b58a8d56be0d6f9f5b10b&vid=e7373bcd667fba9afe3b58a8d56be0d6f9f5b10b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR