4-Pentyn-1-ol ≥96.0%

Supplier: TCI America
Warning

Synonyms: 1-Pentyn-5-ol

P0817-5ML P0817-25ML
TCP0817-005MLEA 50.56 CAD
TCP0817-005ML TCP0817-025ML
4-Pentyn-1-ol ≥96.0%
4-Pentyn-1-ol

CAS Number: 5390-04-5
MDL Number: MFCD00002974
Molecular Formula: C5H8O
Molecular Weight: 84.12
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 155
Flash Point (°C): 62
Specific Gravity (20/20): 0.91

Formula: C₅H₈O
Boiling Pt: 155 °C
Density: 0.91 (20 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00002974
CAS Number: 5390-04-5
EINECS: 226-383-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc1f60fd28668ee3134a980190cd6b6282952066&vid=dc1f60fd28668ee3134a980190cd6b6282952066&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR