Pentaerythrityl tetrabromide ≥98.0%

Supplier: TCI America

Synonyms: 1,3-Dibromo-2,2-bis(bromomethyl)propane, Tetrakis(bromomethyl)methane, Pentaerythrityltetrabromide

P0752-25G P0752-5G
TCP0752-025GEA 295.93 CAD
TCP0752-025G TCP0752-5G
Pentaerythrityl tetrabromide ≥98.0%
Pentaerythrityl tetrabromide

CAS Number: 3229-00-3
MDL Number: MFCD00000210
Molecular Formula: C5H8Br4
Molecular Weight: 387.74
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 306
Melting point (°C): 161


Other Product Names / Synonyms: 1,3-Dibromo-2,2-bis(bromomethyl)propane, Tetrakis(bromomethyl)methane

Formula: C₅H₈Br₄
Boiling Pt: 306 °C
Melting Pt: 161 °C
Density: 2.796 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000210
CAS Number: 3229-00-3
EINECS: 221-764-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e27072073d3094a04be3fc039ac188c81206ff15&vid=5da4cd5db7285a2f044327745f733af0695a5b0c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR