4-Phenylpyridine ≥98.0%

Supplier: TCI America
Warning

P0162-5G P0162-25G
TCP0162-005GEA 86.9 CAD
TCP0162-005G TCP0162-025G
4-Phenylpyridine ≥98.0%
4-Phenylpyridine

CAS Number: 939-23-1
MDL Number: MFCD00006420
Molecular Formula: C11H9N
Molecular Weight: 155.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 281
Melting point (°C): 75

Formula: C₁₁H₉N
Boiling Pt: 281 °C
Melting Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00006420
CAS Number: 939-23-1
EINECS: 213-357-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84ab4c58cafdb276f3a344d3329e64064bdf5c7c&vid=84ab4c58cafdb276f3a344d3329e64064bdf5c7c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR