Phenoxyacetone ≥97.0%

Supplier: TCI America

P0112-5G P0112-25G
TCP0112-5GEA 206.98 CAD
TCP0112-5G TCP0112-025G
Phenoxyacetone ≥97.0%
Phenoxyacetone

CAS Number: 621-87-4
MDL Number: MFCD00008767
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 230
Flash Point (°C): 85
Specific Gravity (20/20): 1.09

Formula: (C₆H₅)OCH₂C(O)CH₃
Boiling Pt: 230 °C
Density: 1.09 (20 °C)
Flash Pt: 85 °C
MDL Number: MFCD00008767
CAS Number: 621-87-4
EINECS: 210-712-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c25323102cf70ac168ee1f4a571f899f16519c88&vid=e154b3813c340760c370cb52fde7b854c1622dd3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR