Octadecylbenzene ≥95.0%
Supplier: TCI America
O0160-25G
O0160-5G
TCO0160-025GEA
601.98
CAD
TCO0160-025G
TCO0160-5G
Octadecylbenzene ≥95.0%
Octadecylbenzene
CAS Number: 4445-07-2
MDL Number: MFCD00048500
Molecular Formula: C24H42
Molecular Weight: 330.60
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Freezing point (°C): 33
Other Product Names / Synonyms: 1-Phenyloctadecane, Stearylbenzene
Formula:
C₂₄H₄₂ Boiling Pt: 408 °C (1013 hPa) Melting Pt: 29 °C Density: 0.8546 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00048500 CAS Number: 4445-07-2 EINECS: 224-684-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...