(S)-(+)-2-Octanol ≥98.0%

Supplier: TCI America
Warning

Synonyms: D(+)-2-Octanol, (S)-Octan-2-ol, (S)-(+)-Hexyl methyl carbinol, (S)-(+)-2-Hydroxy octane, (S)-(+)-Methyl hexyl carbinol

O0144-5ML O0144-25ML
TCO0144-005MLEA 172.22 CAD
TCO0144-005ML TCO0144-25ML
(S)-(+)-2-Octanol ≥98.0%
(S)-(+)-2-Octanol

CAS Number: 6169-06-8
MDL Number: MFCD00064283
Molecular Formula: C8H18O
Molecular Weight: 130.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 76
Specific Gravity (20/20): 0.82
Specific rotation [a]20/D: 10 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: (S)-(+)-Hexylmethylcarbinol

Formula: C₈H₁₈O
MW: 130.23 g/mol
Boiling Pt: 175 °C (1013 hPa)
Density: 0.82 (20 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00064283
CAS Number: 6169-06-8
EINECS: 228-213-6
UN: 1987
ADR: 3,III
Merck Index: 12,06850
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2fce5c428c2c2e21cbab18aca9325eb17a19734d&vid=2fce5c428c2c2e21cbab18aca9325eb17a19734d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR