Ethyl oxamate ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: Oxamic Acid Ethyl Ester, Ethyl 2-amino-2-oxoacetate

O0085-250G O0085-25G
TCO0085-250GEA 211.72 CAD
TCO0085-250G TCO0085-025G
Ethyl oxamate ≥98.0% (by total nitrogen basis)
Ethyl oxamate

CAS Number: 617-36-7
MDL Number: MFCD00008005
Molecular Formula: C4H7NO3
Molecular Weight: 117.10
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 114


Other Product Names / Synonyms: Oxamidic Acid Ethyl Ester, Oxamic Acid Ethyl Ester

Formula: C₄H₇NO₃
MW: 117.1 g/mol
Melting Pt: 114 °C
MDL Number: MFCD00008005
CAS Number: 617-36-7
EINECS: 210-512-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a21e6e4c2197c53b5b9169dad45ccabc6d6722d&vid=4a21e6e4c2197c53b5b9169dad45ccabc6d6722d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR