Dibutyl oxalate ≥99.0%

Supplier: TCI America

O0077-25ML O0077-500ML
TCO0077-025MLEA 48.98 CAD
TCO0077-025ML TCO0077-500ML
Dibutyl oxalate ≥99.0%
Dibutyl oxalate

CAS Number: 2050-60-4
MDL Number: MFCD00009442
Molecular Formula: C10H18O4
Molecular Weight: 202.25
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 246
Melting point (°C): -29
Flash Point (°C): 122
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: Oxalic Acid Dibutyl Ester

Formula: C₁₀H₁₈O₄
Boiling Pt: 246 °C
Melting Pt: –29 °C
Density: 0.99 (20 °C)
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00009442
CAS Number: 2050-60-4
EINECS: 218-092-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5df34b33dfb5687fcfb37123164dbcd717f6bdb&vid=f5df34b33dfb5687fcfb37123164dbcd717f6bdb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR