Sodium oleate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

O0057-25G O0057-500G O0057-100G
TCO0057-025GEA 98.02 CAD
TCO0057-025G TCO0057-500G TCO0057-100G
Sodium oleate ≥97.0% (by titrimetric analysis)
Sodium oleate

CAS Number: 143-19-1
MDL Number: MFCD00004438
Molecular Formula: C18H34O2
Molecular Weight: 304.45
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 235


Product Categories: Materials Chemistry, Anionic Surfactants, Bioscience, Lipids
Other Product Names / Synonyms: Oleic Acid Sodium Salt

Formula: C₁₈H₃₃NaO₂
Melting Pt: 235 °C
Density: 1.02 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00004438
CAS Number: 143-19-1
EINECS: 205-591-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b92ab09173103a6f73278b8d486c0ea4337bf68&vid=3b92ab09173103a6f73278b8d486c0ea4337bf68&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR