4-Nitro-2,1,3-benzothiadiazole ≥98.0%

Supplier: TCI America

Synonyms: 4-Nitrobenzo[c][1,2,5]thiadiazole

N0786-25G N0786-5G
TCN0786-25GEA 374.46 CAD
TCN0786-25G TCN0786-5G
4-Nitro-2,1,3-benzothiadiazole ≥98.0%
4-Nitro-2,1,3-benzothiadiazole

CAS Number: 6583-06-8
MDL Number: MFCD00068050
Molecular Formula: C6H3N3O2S
Molecular Weight: 181.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 109


Other Product Names / Synonyms: 4-Nitropiazthiole

Formula: C₆H₃N₃O₂S
MW: 181.17 g/mol
Melting Pt: 109 °C
MDL Number: MFCD00068050
CAS Number: 6583-06-8
EINECS: 229-514-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a2315e7b7c21cc50d6445fefc78213c0472b418&vid=1a2315e7b7c21cc50d6445fefc78213c0472b418&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR