4-Nitro-1-naphthol ≥98.0%
Supplier: TCI America
Synonyms:
4-Nitronaphthalen-1-ol, KAT8-IN-1, 1-Hydroxy-4-nitronaphthalene
N0672-5G
N0672-25G
TCN0672-5GEA
295.46
CAD
TCN0672-5G
TCN0672-25G
4-Nitro-1-naphthol ≥98.0%
4-Nitro-1-naphthol
CAS Number: 605-62-9
MDL Number: MFCD02179392
Molecular Formula: C10H7NO3
Molecular Weight: 189.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 167
Other Product Names / Synonyms: 1-Hydroxy-4-nitronaphthalene
Formula:
C₁₀H₇NO₃ MW: 189.17 g/mol Melting Pt: 167 °C |
MDL Number:
MFCD02179392 CAS Number: 605-62-9 UN: 2811 ADR: 6.1 I |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...