10-Nonadecanone ≥97.0%

Supplier: TCI America

Synonyms: Caprinone, Dinonyl ketone

N0658-5G N0658-25G
TCN0658-5GEA 189.6 CAD
TCN0658-5G TCN0658-25G
10-Nonadecanone ≥97.0%
10-Nonadecanone

CAS Number: 504-57-4
MDL Number: MFCD00009584
Molecular Formula: C19H38O
Molecular Weight: 282.51
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 156
Melting point (°C): 58
Flash Point (°C): 110


Other Product Names / Synonyms: Caprinone, Dinonyl Ketone

Formula: C₁₉H₃₈O
MW: 282.51 g/mol
Boiling Pt: 156 °C
Melting Pt: 58 °C
Density: 0.806 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009584
CAS Number: 504-57-4
EINECS: 207-994-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72d00eaff15c0cdfae5f86e28d1985ecd212926f&vid=1b7fc4b04453d4d9ebd7fb83fc3abbb492a31b9a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR