(+)-Neomenthol ≥96.0%

Supplier: TCI America

N0626-5ML N0626-25ML
TCN0626-005MLEA 248.8 CAD
TCN0626-005ML TCN0626-025ML
(+)-Neomenthol ≥96.0%
(+)-Neomenthol

CAS Number: 2216-52-6
MDL Number: MFCD00062980
Molecular Formula: C10H20O
Molecular Weight: 156.27
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 212
Flash Point (°C): 82
Specific Gravity (20/20): 0.90
Specific rotation [a]20/D: 17.5 deg (neat)


Product Categories: Bioscience, Monocyclic Monoterpenes
Other Product Names / Synonyms: (1S,2S,5R)-2-Isopropyl-5-methylcyclohexanol

Formula: C₁₀H₂₀O
Boiling Pt: 212 °C
Density: 0.90 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00062980
CAS Number: 2216-52-6
EINECS: 218-691-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3dc0314c46f3df98fb2900faa47e51d8c1453c93&vid=3dc0314c46f3df98fb2900faa47e51d8c1453c93&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR