4-Nitro-p-terphenyl ≥95.0%

Supplier: TCI America

N0610-1G N0610-5G
TCN0610-001GEA 44.24 CAD
TCN0610-001G TCN0610-005G
4-Nitro-p-terphenyl ≥95.0%
4-Nitro-p-terphenyl

CAS Number: 10355-53-0
MDL Number: MFCD00044846
Molecular Formula: C18H13NO2
Molecular Weight: 275.31
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 214

Formula: C₁₈H₁₃NO₂
Melting Pt: 214 °C
MDL Number: MFCD00044846
CAS Number: 10355-53-0
EINECS: 233-775-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=899cf68f85068530bacc357d2fa94c6ec69f1a2c&vid=899cf68f85068530bacc357d2fa94c6ec69f1a2c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR