2,3-Naphthalenedicarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

N0526-5G N0526-25G
TCN0526-005GEA 213.84 CAD
TCN0526-005G TCN0526-25G
2,3-Naphthalenedicarboxylic acid ≥98.0% (by GC, titration analysis)
2,3-Naphthalenedicarboxylic acid

CAS Number: 2169-87-1
MDL Number: MFCD00004104
Molecular Formula: C12H8O4
Molecular Weight: 216.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 246

Formula: C₁₂H₈O₄
Boiling Pt: 422 °C (1013 hPa)
Melting Pt: 246 °C
Storage Temperature: Ambient
MDL Number: MFCD00004104
CAS Number: 2169-87-1
EINECS: 218-517-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85dcde2454c527bae1c11ac2f7394c92a591dcac&vid=85dcde2454c527bae1c11ac2f7394c92a591dcac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR