4-Nitrophthalonitrile ≥98.0%

Supplier: TCI America
Warning

N0524-25G N0524-100G
TCN0524-025GEA 129.88 CAD
TCN0524-025G TCN0524-100G
4-Nitrophthalonitrile ≥98.0%
4-Nitrophthalonitrile

CAS Number: 31643-49-9
MDL Number: MFCD00040301
Molecular Formula: C8H3N3O2
Molecular Weight: 173.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 142


Product Categories: Materials Chemistry, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: 1,2-Dicyano-4-nitrobenzene

Formula: C₈H₃N₃O₂
Melting Pt: 142 °C
MDL Number: MFCD00040301
CAS Number: 31643-49-9
EINECS: 250-748-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc98fcee46312eaa3b226d60d1b92f25997f5e31&vid=cc98fcee46312eaa3b226d60d1b92f25997f5e31&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR