4-Nitroanthranilic acid ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
N0522-25G
N0522-5G
TCN0522-025GEA
37.92
CAD
TCN0522-025G
TCN0522-005G
4-Nitroanthranilic acid ≥97.0% (by HPLC, titration analysis)
4-Nitroanthranilic acid
CAS Number: 619-17-0
MDL Number: MFCD00007262
Molecular Formula: C7H6N2O4
Molecular Weight: 182.14
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 262
Other Product Names / Synonyms: 2-Amino-4-nitrobenzoic Acid
Formula:
C₇H₆N₂O₄ Melting Pt: 262 °C |
MDL Number:
MFCD00007262 CAS Number: 619-17-0 EINECS: 210-583-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...