Norbornane ≥98.0%
Supplier: TCI America
N0517-10G
N0517-25G
TCN0517-010GEA
309.68
CAD
TCN0517-010G
TCN0517-25G
Norbornane ≥98.0%
Norbornane
CAS Number: 279-23-2
MDL Number: MFCD00074822
Molecular Formula: C7H12
Molecular Weight: 96.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 106
Melting point (°C): 85
Other Product Names / Synonyms: Bicyclo[2.2.1]heptane
Formula:
C₇H₁₂ |
MDL Number:
MFCD00074822 CAS Number: 279-23-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...