1,8-Naphthyridine ≥98.0%
Supplier: TCI America
Synonyms:
1,8-Diazanaphthalene, Pyrido[2,3-b]pyridine
N0464-100MG
N0464-1G
TCN0464-100MGEA
41.08
CAD
TCN0464-100MG
TCN0464-001G
1,8-Naphthyridine ≥98.0%
1,8-Naphthyridine
CAS Number: 254-60-4
MDL Number: MFCD00059751
Molecular Formula: C8H6N2
Molecular Weight: 130.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 98
Other Product Names / Synonyms: 1,8-Diazanaphthalene, Pyrido[2,3-b]pyridine
Formula:
C₈H₆N₂ MW: 130.1466 g/mol Melting Pt: 98 °C |
MDL Number:
MFCD00059751 CAS Number: 254-60-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...