Neocuproine hydrochloride monohydrate ≥99.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
2,9-Dimethyl-1,10-phenanthroline hydrochloride
N0423-1G
N0423-5G
TCN0423-001GEA
64.78
CAD
TCN0423-001G
TCN0423-5G
Neocuproine hydrochloride monohydrate ≥99.0% (by HPLC, titration analysis)
Neocuproine hydrochloride monohydrate
CAS Number: 303136-82-5
MDL Number: MFCD00012645
Molecular Formula: C14H12N2
Molecular Weight: 244.72
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Product Categories: Synthetic Organic Chemistry, Ligands, Analytical Science, Phenanthrolines (Chelating Agents)
Other Product Names / Synonyms: 2,9-Dimethyl-1,10-phenanthroline Hydrochloride
Formula:
C₁₄H₁₃ClN₂·1H₂O MW: 262.74 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00012645 CAS Number: 303136-82-5 EINECS: 230-732-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...