1,5-Naphthyridine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

N0401-1G N0401-100MG
TCN0401-001GEA 181.7 CAD
TCN0401-001G TCN0401-100MG
1,5-Naphthyridine ≥98.0% (by titrimetric analysis)
1,5-Naphthyridine

CAS Number: 254-79-5
MDL Number: MFCD00039727
Molecular Formula: C8H6N2
Molecular Weight: 130.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 152
Melting point (°C): 74


Other Product Names / Synonyms: 1,5-Diazanaphthalene, Pyrido[1,5-b]pyridine

Formula: C₈H₆N₂
Boiling Pt: 152 °C
Melting Pt: 74 °C
MDL Number: MFCD00039727
CAS Number: 254-79-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9173e21d9c4e11a46e9230841702cdde66e1029a&vid=9173e21d9c4e11a46e9230841702cdde66e1029a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR