Nonylcyclohexane ≥98.0%

Supplier: TCI America

N0379-5ML N0379-25ML
TCN0379-005MLEA 158.63 CAD
TCN0379-005ML TCN0379-025ML
Nonylcyclohexane ≥98.0%
Nonylcyclohexane

CAS Number: 2883-02-5
MDL Number: MFCD00058955
Molecular Formula: C15H30
Molecular Weight: 210.41
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 282
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: 1-Cyclohexylnonane

Formula: C₁₅H₃₀
Boiling Pt: 282 °C
Melting Pt: –10 °C
Density: 0.82 (20 °C)
MDL Number: MFCD00058955
CAS Number: 2883-02-5
EINECS: 220-739-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceb8dbaea1331e29bcea9018832fba75190ed68c&vid=ceb8dbaea1331e29bcea9018832fba75190ed68c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR