4-Nitrocinnamic acid ≥99.0%

Supplier: TCI America

Synonyms: 3-(4-Nitrophenyl)propenoic acid, 3-(4-Nitrophenyl)acrylic acid

N0355-25G N0355-10G
TCN0355-025GEA 195.92 CAD
TCN0355-025G TCN0355-010G
4-Nitrocinnamic acid ≥99.0%
4-Nitrocinnamic acid

CAS Number: 619-89-6
MDL Number: MFCD00007381
Molecular Formula: C9H7NO4
Molecular Weight: 193.16
Purity/Analysis Method: >99.0% (T)
Form: Crystal

Formula: C₉H₇NO₄
MDL Number: MFCD00007381
CAS Number: 619-89-6
EINECS: 212-924-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4681eb455cd9be310ec3847e4469736bafd11c4&vid=b9d81111fb9cabe8acf994fca10681defe5e0175&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR