1-Nonadecanol ≥98.0%

Supplier: TCI America

Synonyms: Nonadecyl alcohol

N0284-5G N0284-25G
TCN0284-005GEA 246.48 CAD
TCN0284-005G TCN0284-025G
1-Nonadecanol ≥98.0%
1-Nonadecanol

CAS Number: 1454-84-8
MDL Number: MFCD00002824
Molecular Formula: C19H40O
Molecular Weight: 284.53
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 62


Product Categories: Bioscience, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Nonadecyl Alcohol

Formula: H₃C(CH₂)₁₇CH₂OH
MW: 284.53 g/mol
Boiling Pt: 345 °C (1013 hPa)
Melting Pt: 62 °C
Density: 0.882 g/cm³ (21 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002824
CAS Number: 1454-84-8
EINECS: 215-930-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f1e6f95c175934fdcb4a90f55aad1fc71a2e8cb&vid=373d830fa5608fb0469d0a943564bf6c0b0787eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR