2-Nitrobiphenyl ≥98.0%

Supplier: TCI America

N0194-25G N0194-500G
TCN0194-025GEA 37.92 CAD
TCN0194-025G TCN0194-500G
2-Nitrobiphenyl ≥98.0%
2-Nitrobiphenyl

CAS Number: 86-00-0
MDL Number: MFCD00007126
Molecular Formula: C12H9NO2
Molecular Weight: 199.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 170
Freezing point (°C): 36

Formula: C₁₂H₉NO₂
MW: 199.21 g/mol
Boiling Pt: 320
Melting Pt: 36
Density: 1.44 g/cm³ (20 °C)
MDL Number: MFCD00007126
CAS Number: 86-00-0
EINECS: 201-646-8
Merck Index: 12,06689
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55755293fc9731b83d6a44ea072c6b970f290ecf&vid=55755293fc9731b83d6a44ea072c6b970f290ecf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR