Nitrilotriacetic acid disodium salt ≥99.0%
Supplier: TCI America
Synonyms:
Nitrilotriacetic acid, disodium salt
N0100-25G
N0100-500G
TCN0100-025GEA
67.94
CAD
TCN0100-025G
TCN0100-500G
Nitrilotriacetic acid disodium salt ≥99.0%
Nitrilotriacetic acid disodium salt
CAS Number: 15467-20-6
MDL Number: MFCD00064230
Molecular Formula: C6H9NO6
Molecular Weight: 235.10
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Product Categories: Analytical Science, Complexones (Chelating Agents)
Other Product Names / Synonyms: Triglycollamic Acid Disodium Salt, Disodium Nitrilotriacetate, Disodium Hydrogen Nitrilotriacetate, NTA 2Na
Formula:
C₆H₇NNa₂O₆ MW: 235.1 g/mol |
MDL Number:
MFCD00064230 CAS Number: 15467-20-6 EINECS: 239-484-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...