1,2-Naphthoquinone ≥95.0% (by HPLC and gravimetric analysis)

Supplier: TCI America
Warning

N0041-5G N0041-1G
TCN0041-005GEA 187.91 CAD
TCN0041-005G 76220-350
1,2-Naphthoquinone ≥95.0% (by HPLC and gravimetric analysis)
1,2-Naphthoquinone

CAS Number: 524-42-5
MDL Number: MFCD00001698
Molecular Formula: C10H6O2
Molecular Weight: 158.16
Purity/Analysis Method: >95.0% (HPLC,W)
Form: Crystal
Melting point (°C): 133
Storage Temperature: 0-10°C

Formula: C₁₀H₆O₂
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 133 °C
Density: 1.45 g/cm³ (20 °C)
MDL Number: MFCD00001698
CAS Number: 524-42-5
EINECS: 208-360-2
Merck Index: 12,06481
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73354635a7240f451f4498790de6173455d58703&vid=73354635a7240f451f4498790de6173455d58703&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,W)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR