2-Naphthoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

N0025-25G N0025-100G N0025-5G
TCN0025-025GEA 66.36 CAD
TCN0025-025G TCN0025-100G TCN0025-5G
2-Naphthoic acid ≥98.0% (by GC, titration analysis)
2-Naphthoic acid

CAS Number: 93-09-4
MDL Number: MFCD00004101
Molecular Formula: C11H8O2
Molecular Weight: 172.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 185
Flash Point (°C): 205


Product Categories: Analytical Science, Absolute Configuration Determination (Exciton Chirality CD Method)
Other Product Names / Synonyms: 2-Naphthalenecarboxylic Acid

Formula: C₁₁H₈O₂
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 185 °C
Density: 1.382 g/cm³ (20 °C)
Flash Pt: 205 °C
MDL Number: MFCD00004101
CAS Number: 93-09-4
EINECS: 202-217-8
Merck Index: 12,06469
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=877b9e057d22e1ce9a92282c138a0aa9c3762a4d&vid=877b9e057d22e1ce9a92282c138a0aa9c3762a4d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR