Naphthacene ≥97.0% (by HPLC)
Supplier: TCI America
N0001-1G
N0001-100MG
N0001-5G
TCN0001-001GEA
404.48
CAD
TCN0001-001G
TCN0001-100MG
TCN0001-5G
Naphthacene ≥97.0% (by HPLC)
Naphthacene
CAS Number: 92-24-0
MDL Number: MFCD00003702
Molecular Formula: C18H12
Molecular Weight: 228.29
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 357
Product Categories: Materials Chemistry, Organic Transistor Materials, Solar Cell Research Reagents, Nanocarbon Unit Structures (Nanocarbon Materials)
Other Product Names / Synonyms: 2,3-Benzanthracene, Tetracene
Formula:
C₁₈H₁₂ Boiling Pt: 437 °C (1013 hPa) Melting Pt: 357 °C Density: 1.323 g/cm³ (20 °C) |
MDL Number:
MFCD00003702 CAS Number: 92-24-0 EINECS: 202-138-9 Merck Index: 12,06456 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...