3-Methyl-5-(trifluoromethyl)-1H-pyrazole ≥98.0%

Supplier: TCI America

Synonyms: 3-Methyl-5-(trifluoromethyl)pyrazole

M2032-1G M2032-5G
TCM2032-1GEA 61.13 CAD
TCM2032-1G TCM2032-5G
3-Methyl-5-(trifluoromethyl)-1H-pyrazole ≥98.0%
3-Methyl-5-(trifluoromethyl)-1H-pyrazole

CAS Number: 10010-93-2
MDL Number: MFCD00156058
Molecular Formula: C5H5F3N2
Molecular Weight: 150.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 89

Formula: C₅H₅F₃N₂
MW: 150.104 g/mol
Melting Pt: 89 °C
MDL Number: MFCD00829309
CAS Number: 10010-93-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64803848712d13307c8072f15948f7c1f0823593&vid=64803848712d13307c8072f15948f7c1f0823593&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR